Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB320841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320841 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320841 Supplier Novus Biologicals Supplier No. NBP298357JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEK3 Polyclonal antibody specifically detects NEK3 in Mouse samples. It is validated for ELISA

Specifications

Antigen NEK3
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, glycogen synthase A kinase, HSPK 36, HSPK36, hydroxyalkyl-protein kinase, MGC29949, Never in mitosis A-related kinase 3, NIMA (never in mitosis gene a)-related kinase 3, NimA-related protein kinase 3, phosphorylase B kinase kinase, serine/threonine-protein kinase Nek3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse NEK3 (Accession#: ABK42288.1; Met1-Ala509)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4752
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.