Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK6 Antibody (OTI5D7), PerCP, Novus Biologicals™
SDP

Catalog No. NB272938PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272938PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272938PCP Supplier Novus Biologicals Supplier No. NBP272938PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK6 Monoclonal specifically detects NEK6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen NEK6
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5D7
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, Never in mitosis A-related kinase 6, NIMA (never in mitosis gene a)-related kinase 6, NimA-related protein kinase 6, Protein kinase SID6-1512, putative serine-threonine protein kinase, serine/threonine-protein kinase Nek6, SID6-1512
Gene Symbols NEK6
Host Species Mouse
Immunogen Full length recombinant protein of human NEK6 (NP_055212) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10783.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.