Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK9 Antibody (OTI1H1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272939IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272939IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272939IR Supplier Novus Biologicals Supplier No. NBP272939IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NEK9 Monoclonal specifically detects NEK9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NEK9
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H1
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp434D0935, KIAA1995, MGC138306, MGC16714, NEK8, Nek8NERCC1, Nercc1 kinase, NERCCEC 2.7.11.1, Never in mitosis A-related kinase 9, NIMA (never in mitosis gene a)- related kinase 9, NimA-related kinase 8, NIMA-related kinase Nek8, NimA-related protein kinase 9, serine/threonine-protein kinase Nek9
Gene Symbols NEK9
Host Species Mouse
Immunogen Full length recombinant protein of human NEK9 (NP_149107) produced in HEK293 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, DNA Repair, GPCR, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 91754.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.