Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Nelfinavir mesylate

Click to view available options
Quantity:
10 mg
50 mg
Description
Orally active human immunodeficiency virus protease inhibitor. Potently inhibits HIV-1 protease (Ki = 2 nM) in vitro. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.08 μM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 663.89 |
Purity | >98% |
Molecular Weight (g/mol) | 663.889 |
Chemical Name or Material | Nelfinavir mesylate |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction