Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NELL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219511 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219511 Supplier Novus Biologicals Supplier No. NBP219511

Rabbit Polyclonal Antibody

NELL1 Polyclonal specifically detects NELL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NELL1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.85mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. Q92832
Gene Alias FLJ45906, IDH3GL, nel (chicken)-like 1, NEL-like 1 (chicken), NEL-like protein 1, Nel-related protein 1, neural epidermal growth factor-like 1, NRP1, protein kinase C-binding protein NELL1
Gene Symbols NELL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NELL1. The exact sequence is proprietary.
Molecular Weight of Antigen 90 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4745
Test Specificity Human. Species reactivity based on sequence identity: Mouse (95%), Rat (94%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.