Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NELL2 Antibody (055), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB070684 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070684 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070684 Supplier Novus Biologicals Supplier No. NBP290179G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NELL2 Monoclonal specifically detects NELL2 in Human samples. It is validated for ELISA.

Specifications

Antigen NELL2
Applications ELISA
Classification Monoclonal
Clone 55
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human NELL2 (Q99435; Met1-Leu816).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4753
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.