Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NELL2 Antibody (13E7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP174039A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP174039A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP174039A30 Supplier Novus Biologicals Supplier No. NBP174039AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NELL2 Monoclonal specifically detects NELL2 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen NELL2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 130000000
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2
Gene Symbols NELL2
Host Species Mouse
Immunogen Recombinant human NELL2 (30-258aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4753.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.