Learn More
NEMF Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | NEMF |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | NEMF |
| Gene Accession No. | O60524 |
| Gene Alias | Nuclear export mediator factor NEMF, Antigen NY-CO-1, Serologically defined colon cancer antigen 1, NEMF, SDCCAG1 |
| Gene Symbols | NEMF |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.