Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nemonapride
GREENER_CHOICE

Catalog No. 174610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-461-0 10 mg
17-465-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-461-0 Supplier Tocris Bioscience™ Supplier No. 1746/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent D2-like antagonist. Also 5-HT1A agonist

Highly potent dopamine D2-like receptor antagonist; selective over D1-like receptors (Ki values are 0.1 and 740 nM for D2-like and D1-like receptors respectively). Also potent 5-HT1A receptor agonist (IC50 = 34 nM) and has affinity for sigma receptors.

Chemical Identifiers

CAS 75272-39-8
Molecular Formula C21H26ClN3O2

Specifications

Quantity 10 mg
Formula Weight 387.91
Purity >99%
Molecular Weight (g/mol) 387.908
Chemical Name or Material Nemonapride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.