Learn More
Neodymium(III) carbonate hydrate, 99.9% (REO)
Description
Neodymium Carbonate mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 50 g |
| Solubility Information | Insoluble in water,moderately soluble in strong mineral acids. |
| Formula Weight | 468.51 (Anhydrous) |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Chemical Name or Material | Neodymium(III) carbonate hydrate |
Safety and Handling
| EINECSNumber | 227-579-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.