Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) isopropoxide, 99% (REO), 2.5% w/v in toluene/isopropanol

Catalog No. AA36573AC
Click to view available options
Quantity:
25 mL
100 mL

Catalog No. AA36573AC

Supplier: Thermo Scientific Chemicals 036573AC

Only null left
Add to Cart

CAS: 19236-15-8 | C9H24NdO3 | 324.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19236-15-8
Molecular Formula C9H24NdO3
Molecular Weight (g/mol) 324.53
MDL Number MFCD00058819,MFCD00058819
InChI Key WOCLHTSADRFWMF-UHFFFAOYSA-N
Synonym neodymium i-propoxide, neodymium iii isopropoxide
PubChem CID 25199945
SMILES [Nd].CC(C)O.CC(C)O.CC(C)O

Specifications

Color Blue
Linear Formula Nd[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
IUPAC Name tris(propan-2-ol) neodymium
Formula Weight 321.3
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 2.5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Neodymium(III) isopropoxide
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H318-H336-H315
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.