Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) nitride, 99.9% (REO)

Catalog No. aa4209104
Click to view available options
Quantity:
2 g
10 g

CAS: 25764-11-8 | NNd | 158.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25764-11-8
Molecular Formula NNd
Molecular Weight (g/mol) 158.249
MDL Number MFCD00049990
InChI Key OVMJQLNJCSIJCH-UHFFFAOYSA-N
Synonym neodymium nitride, neodymium nitride ndn, neodymium iii nitride, neodymiumylidyneamine
PubChem CID 117628
IUPAC Name azanylidyneneodymium
SMILES N#[Nd]

Specifications

Physical Form Powder
Quantity 2 g
Formula Weight 158.25
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium(III) nitride
EINECSNumber 247-246-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.