Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neogenin Antibody (RM0124-3G55), HRP, Novus Biologicals™
SDP

Catalog No. NBP122501H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122501H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122501H Supplier Novus Biologicals Supplier No. NBP122501H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Neogenin Monoclonal specifically detects Neogenin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Neogenin
Applications Western Blot
Classification Monoclonal
Clone RM0124-3G55
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias HsT17534, IGDCC2DKFZp547B146, Immunoglobulin superfamily DCC subclass member 2, neogenin, neogenin (chicken) homolog 1, neogenin 1, neogenin homolog 1, NGNDKFZp547A066
Gene Symbols NEO1
Host Species Rat
Immunogen Mouse recombinant protein of Neogenin
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4756
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of Neogenin.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.