Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nephrin Antibody (Y17-R), Novus Biologicals™
SDP

Catalog No. NBP13013005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP13013005 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP13013005 Supplier Novus Biologicals Supplier No. NBP1301300.05ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody has been used in 6 publications

Nephrin Monoclonal specifically detects Nephrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Nephrin
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone Y17-R
Concentration 4.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000, ELISA 1:50000-1:100000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500
Formulation 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. O60500, Q9QZS7, Q9R044
Gene Alias nephrin, nephrosis 1, congenital, Finnish type (nephrin), NPHNCNF, Renal glomerulus-specific cell adhesion receptor
Gene Symbols NPHS1
Host Species Rabbit
Immunogen Peptide derived from the centre of the fifth Ig-like domain of human nephrin (between residues Cys465-Cys528).
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4868
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.