Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nephrin Monoclonal Antibody (C8)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Nephrin |
---|---|
Clone | C8 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot, Immunocytochemistry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541644
|
Invitrogen™
MA541644 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543718
|
Invitrogen™
MA543718 |
20 μL |
Each for $213.50
|
|
|||||
Description
Nephrin Monoclonal Antibody for Western Blot, ICC/IF
Primary steroid resistant nephrotic syndrome (SRNS) is characterized by childhood onset of proteinuria and progression to end stage renal disease. Kidney podocytes and their slit diaphragms form the final barrier to urinary protein loss. Congenital nephrotic syndrome (CNS) is caused by mutations in NPHS1 (nephrin) or NPHS2. Nephrin, a recently identified protein is a member of a group of podocyte proteins that constitute major component of the slit diaphragm especially in the foot process. Nephrin, a cell adhesion molecule, may play a crucial role in maintaining the glomerular filtration barrier. Recent studies have suggested that mutations in the gene for Nephrin reportedly lead to congenital nephrosis. Three novel podocyte proteins, Podocin, Nephrin and alpha Actinin 4 have been identified in congenital and experimental models of proteinuria. The role of Nephrin in anti apoptotic activity in podocyte slit diaphragm is believed to be associated with vascular endothelial derived growth factors VEGF signaling. Nephrin seems to play a role in the development or function of the kidney glomerular filtration barrier and it may anchor the podocyte slit diaphragm to the actin cytoskeleton.Specifications
Nephrin | |
1 mg/mL | |
Western Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O60500 | |
4868 | |
Recombinant protein Nephrin. The antigen corresponds to amino acid range 27-234 of the target protein. | |
Antibody |
C8 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
Nphs1 | |
CNF; Nephrin; nephrin 1; NephrinB; nephrosis 1 homolog, nephrin; nephrosis 1, congenital, Finnish type (nephrin); nephrosis 1, nephrin; NPHN; NPHS1; NPHS1 nephrin; NPHS1, nephrin; renal glomerulus-specific cell adhesion receptor | |
Nphs1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title