Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neprilysin/CD10 Antibody (113), Alexa Fluor™ 647
SDP

Catalog No. NB087998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087998 Supplier Novus Biologicals Supplier No. NBP289226AF647

Rabbit Monoclonal Antibody

Neprilysin/CD10 Monoclonal specifically detects Neprilysin/CD10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Neprilysin/CD10
Applications Immunofluorescence
Classification Monoclonal
Clone 113
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Neprilysin/CD10.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4311
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.