Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neprilysin/CD10 Antibody (E19-P), Novus Biologicals™
SDP

Catalog No. NBP17899610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17899610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17899610 Supplier Novus Biologicals Supplier No. NBP178996100UL

Rabbit Monoclonal Antibody

Neprilysin/CD10 Monoclonal specifically detects Neprilysin/CD10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Neprilysin/CD10
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E19-P
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200
Formulation 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P08473
Gene Alias Atriopeptidase, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11, SFE, Skin fibroblast elastase
Gene Symbols MME
Host Species Rabbit
Immunogen Peptide derived from the C-terminal sequence of human CD10
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4311
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.