Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nesprin 2 Antibody (10H8), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP259945V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-59945V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-59945V Supplier Novus Biologicals Supplier No. NBP259945V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nesprin 2 Monoclonal specifically detects Nesprin 2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nesprin 2
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 10H8
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH
Gene Symbols SYNE2
Host Species Mouse
Immunogen GST fused to residues 6,653-6,835 of human Nesprin 2
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 23224
Test Specificity Specificity of human Nesprin 2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.