Learn More
Nestin Antibody (10C2) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 21 publication
Supplier:  Novus Biologicals NB300266
 View more versions of this product
Catalog No. NB300266
Only null left
Add to Cart
Description
Description
Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
Specifications
Nestin
Monoclonal
2.4 mg/mL
Western Blot 1:1000 - 1:5000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, CyTOF-ready
FLJ21841, nestin
Mouse
Protein G purified
RUO
Primary
Human. It does not cross-react mouse, rat or rabbit.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
10C2
Unconjugated
P48681
NES
A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681]
0.1 mL
Angiogenesis, Cancer, Cancer Stem Cells, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
10763
Human, Mouse, Rabbit (Negative), Rat (Negative)
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only