Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody (10C2) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 21 publication

$257.00 - $574.50

Specifications

Antigen Nestin
Clone 10C2
Concentration 2.4 mg/mL
Dilution Western Blot 1:1000 - 1:5000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300266SS
SDP
View Documents
Novus Biologicals
NB300266SS
0.025 mL
Each for $257.00
Only null left
Add to Cart
 
NB300266
SDP
View Documents
Novus Biologicals
NB300266
0.1 mL
Each for $574.50
Only null left
Add to Cart
 
Description

Description

Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications

Specifications

Nestin
2.4 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Cancer, Cancer Stem Cells, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
P48681
10763
A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
10C2
Western Blot 1:1000 - 1:5000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, CyTOF-ready
Monoclonal
Purified
RUO
Human, Mouse, Rabbit (Negative), Rat (Negative)
FLJ21841, nestin
NES
IgG1 κ
Protein G purified
Human. It does not cross-react mouse, rat or rabbit.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.