Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody (196908), HRP, Novus Biologicals™
SDP

Catalog No. NB350049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350049 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350049 Supplier Novus Biologicals Supplier No. FAB1259H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-reported

Specifications

Antigen Nestin
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 196908
Conjugate HRP
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-reported
Formulation PBS
Gene Alias FLJ21841, nestin
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Nestin
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10763
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.