Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB30265A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30265A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30265A750 Supplier Novus Biologicals Supplier No. NB300265AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nestin Polyclonal specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nestin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ21841, nestin
Gene Symbols NES
Host Species Rabbit
Immunogen Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProtNo. P48681]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cancer Stem Cells, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10763.0
Test Specificity This is specific for human nestin.
Target Species Human, Canine, Mouse, Rabbit, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.