Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB300265B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300265B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300265B Supplier Novus Biologicals Supplier No. NB300265B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nestin Polyclonal specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nestin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P48681
Gene Alias FLJ21841, nestin
Gene Symbols NES
Host Species Rabbit
Immunogen Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProt# P48681]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10763
Test Specificity This is specific for human nestin.
Target Species Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.