Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nestin Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB359434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359434 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359434 Supplier Novus Biologicals Supplier No. NB300265MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nestin Polyclonal antibody specifically detects Nestin in Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Nestin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ21841, nestin
Host Species Rabbit
Immunogen Recombinant human Nestin protein (corresponding to residues 1464-1614). [UniProt# P48681]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10763
Target Species Human, Canine, Mouse (Negative), Rabbit (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.