Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Netrin-G1a Antibody (1010035) [FITC], Novus Biologicals™
SDP

Catalog No. NB402707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402707 Supplier Novus Biologicals Supplier No. FAB10019F

Mouse Monoclonal Antibody

Netrin-G1a Monoclonal antibody specifically detects Netrin-G1a in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Netrin-G1a
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1010035
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Lmnt1, NTNG1
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human Netrin-G1a, His29-Gly503, Accession # Q9Y212
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22854
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.