Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEU2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221649 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221649 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221649 Supplier Novus Biologicals Supplier No. NBP221649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NEU2 Polyclonal specifically detects NEU2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen NEU2
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.9 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000-1:50000, Immunohistochemistry, Immunoprecipitation
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9Y3R4
Gene Alias Cytosolic sialidase, EC 3.2.1.18, N-acetyl-alpha-neuraminidase 2, neuraminidase 2, SIAL2MGC129579, sialidase 2 (cytosolic sialidase), sialidase-2
Gene Symbols NEU2
Host Species Rabbit
Immunogen This affinity purified NEU2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal portion near aa 100-125 of Human NEU2. (Uniprot: Q9Y3R4)
Molecular Weight of Antigen 42.25 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4759
Test Specificity This antibody reacts with human NEU2. Based on sequence we expect this antibody to react with neuraminidase from other sources, although specific reactivity has not been confirmed. Cross-reactivity against Neu1 has not yet been established.
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.