Learn More
NEU3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 278791AP150UL
Description
The NEU3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NEU3. This antibody recognizes human, mouse antigen. The NEU3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NEU3 | |
Polyclonal | |
Unconjugated | |
150 μL | |
10825 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
270 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.