Learn More
NEU3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NEU3 |
---|---|
Concentration | 270 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8591
![]() ![]() |
Proteintech Group Inc
278791AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The NEU3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NEU3. This antibody recognizes human, mouse antigen. The NEU3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
NEU3 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG |
270 μg/mL | |
Polyclonal | |
Rabbit | |
10825 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.