Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuregulin-1/NRG1 Antibody (005), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB071126 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB071126 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB071126 Supplier Novus Biologicals Supplier No. NBP290220AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Neuregulin-1/NRG1 Monoclonal specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for ELISA.

Specifications

Antigen Neuregulin-1/NRG1
Applications ELISA
Classification Monoclonal
Clone 5
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Neuregulin-1/NRG1 (Q02297-1; Ser20-Lys241).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3084
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.