Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuregulin-1/NRG1 Antibody (NRG1/2710), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB391229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391229 Supplier Novus Biologicals Supplier No. NBP308715AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuregulin-1/NRG1 Monoclonal antibody specifically detects Neuregulin-1/NRG1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Neuregulin-1/NRG1
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone NRG1/2710
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1a, MST131, MSTP131, NDFheregulin, alpha (45kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Host Species Mouse
Immunogen Recombinant fragment of human Neuregulin-1/NRG1 protein (around aa 21-242) (exact sequence is proprietary) (Uniprot: Q02297)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3084
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.