Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuregulin-3/NRG3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB081158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081158 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081158 Supplier Novus Biologicals Supplier No. NBP297121R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neuregulin-3/NRG3 Polyclonal specifically detects Neuregulin-3/NRG3 in Human samples. It is validated for ELISA.

Specifications

Antigen Neuregulin-3/NRG3
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias HRG3, neuregulin 3, neuregulin 3 variant 1, neuregulin 3 variant 10, neuregulin 3 variant 11, neuregulin 3 variant 12, neuregulin 3 variant 13, neuregulin 3 variant 14, neuregulin 3 variant 3, neuregulin 3 variant 4, neuregulin 3 variant 5, neuregulin 3 variant 6, neuregulin 3 variant 7, neuregulin 3 variant 8, neuregulin 3 variant 9, neuregulin-3-like polypeptide, pro-neuregulin-3, membrane-bound isoform, pro-NRG3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Neuregulin-3/NRG3 (ADK90021.1; Glu282-Pro342).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10718
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.