Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroblastoma Antibody (FMG25), FITC, Novus Biologicals™
SDP

Catalog No. NBP250528F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250528F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250528F Supplier Novus Biologicals Supplier No. NBP250528F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuroblastoma Monoclonal specifically detects Neuroblastoma in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Neuroblastoma
Applications Flow Cytometry
Classification Monoclonal
Clone FMG25
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Human cutaneous T cell lymphoma (HUT 78), SPF female Balb/c mice were injected i.p with 2 x 10*5 cells in complete Freund's adjuvant. 30 days later a 2nd i.p injection of 2 x 10*5 cells without adjuvant was given. Fusion was 5 days
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.