Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NeuroD1 Rabbit anti-Human, Clone: JM11-10, Novus Biologicals™
SDP

Catalog No. NBP266971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266971 Supplier Novus Biologicals Supplier No. NBP266971

Rabbit Monoclonal Antibody

NeuroD1 Monoclonal specifically detects NeuroD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen NeuroD1
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM11-10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:10-1:50, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias basic helix-loop-helix transcription factor, BETA2, BHF-1, BHLHA3, bHLHa3beta-cell E-box transactivator 2, Class A basic helix-loop-helix protein 3, MODY6, NeuroD, NeuroD1, neurogenic differentiation 1, neurogenic differentiation factor 1, neurogenic helix-loop-helix protein NEUROD, NIDDM
Gene Symbols NEUROD1
Host Species Rabbit
Immunogen Synthetic peptide within Human NeuroD1 aa 1-44/ 356. (SwissProt: Q13562 Human; SwissProt: Q60867 Mouse; SwissProt: Q64289 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 4760
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.