Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Neurodazine
GREENER_CHOICE

Catalog No. 365610R
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Induces neurogenesis in mature skeletal muscle cells

Neurodazine is an induces neurogenesis of non-pluripotent C2C12 myoblasts and cells derived from mature human skeletal muscle. Upregulates the expression of neuron-specific markers in C2C12 cells.

Specifications

Quantity 10 mg
Product Type Neurodazine
Purity >99%
Molecular Weight (g/mol) 456.92
Inhibitors Neurodazine

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.