Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofascin Antibody (1018843) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB373408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373408 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373408 Supplier Novus Biologicals Supplier No. FAB8208A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neurofascin Monoclonal specifically detects Neurofascin in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Neurofascin
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1018843
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NRCAML
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human Neurofascin, Ile25-Trp1039, Accession # NP_055905
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23114
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.