Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofascin Antibody, Novus Biologicals™
SDP

Catalog No. NBP178794 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178794 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178794 Supplier Novus Biologicals Supplier No. NBP178794
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Neurofascin Polyclonal specifically detects Neurofascin in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Neurofascin
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 μg/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001005388.2
Gene Alias DKFZp686P2250, KIAA0756FLJ46866, neurofascin, neurofascin homolog, neurofascin homolog (chicken), NF, NRCAML
Gene Symbols NFASC
Host Species Goat
Immunogen Peptide with sequence C-NFSPNQTKFTVQRT corresponding to internal region according to NP_001153803.1, NP_001153804.1, NP_055905.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23114
Test Specificity This antibody is expected to recognize reported isoforms 1, 2, 3 and 4 (NP_001005388.2;,NP_001153803.1, NP_001153804.1and NP_055905.2 resp) .
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.