Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibromin 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300155SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300155SS 0.125 mL
NB300155 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300155SS Supplier Novus Biologicals Supplier No. NB300155SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Neurofibromin 1 Polyclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Neurofibromin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23910086), Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen 1:200-1:500
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. P21359
Gene Alias DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Gene Symbols NF1
Host Species Rabbit
Immunogen An N-terminal peptide of Neurofibromin 1 (within amino acids 1-50). [Uniprot: P21359]
Purification Method Unpurified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4763
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.