Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibromin 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 4 publications

$207.00 - $501.50

Specifications

Antigen Neurofibromin 1
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23910086), Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen 1:200-1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300155SS
SDP
View Documents
Novus Biologicals
NB300155SS
0.125 mL
Each for $207.00
Only null left
Add to Cart
 
NB300155
SDP
View Documents
Novus Biologicals
NB300155
0.5 mL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

Neurofibromin 1 Polyclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Neurofibromin 1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Whole antisera with 0.02% Sodium Azide
DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
NF1
IgG
Unpurified
Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23910086), Immunohistochemistry-Paraffin 1:200-1:500, Immunohistochemistry-Frozen 1:200-1:500
Polyclonal
Antisera
RUO
Human, Mouse, Rat
P21359
4763
An N-terminal peptide of Neurofibromin 1 (within amino acids 1-50). [Uniprot: P21359]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.