Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofibromin 1 Antibody (McNFn27a) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB300153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300153 0.2 mL
NB300153SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300153 Supplier Novus Biologicals Supplier No. NB300153
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Neurofibromin 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone McNFn27a
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:20 - 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50. Use reported in scientific literature
Gene Accession No. P21359
Gene Alias DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Gene Symbols NF1
Host Species Mouse
Immunogen A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359]
Purification Method Affinity Purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4763
Test Specificity This is specific for full-length or truncated forms of neurofibromin.
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.