Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068155 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068155 Supplier Novus Biologicals Supplier No. NB10068155

Goat Polyclonal Antibody

Neurogenin-1 Polyclonal specifically detects Neurogenin-1 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.

Specifications

Antigen Neurogenin-1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006152.2
Gene Alias AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1
Gene Symbols NEUROG1
Host Species Goat
Immunogen Peptide with sequence KDLLHTTPCFIPYH corresponding to C-Terminus according to NP_006152.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4762
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.