Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093528 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093528 Supplier Novus Biologicals Supplier No. NB10093528
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Neurogenin-1 Polyclonal specifically detects Neurogenin-1 in Zebrafish samples. It is validated for Peptide ELISA.

Specifications

Antigen Neurogenin-1
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_571116.1
Gene Alias AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1
Gene Symbols NEUROG1
Host Species Goat
Immunogen Peptide with sequence C-SHTDDEDSRSSLH corresponding to internal region according to Uniprot Zebrafish NP_571116.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4762
Target Species Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.