Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin-2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB106731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106731 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106731 Supplier Novus Biologicals Supplier No. NBP298917B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neurogenin-2 Polyclonal specifically detects Neurogenin-2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Neurogenin-2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, neurogenin 2, neurogenin-2, ngn-2, NGN2, Protein atonal homolog 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Neurogenin-2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63973
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.