Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Neurogranin Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Neurogranin |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541648
|
Invitrogen™
MA541648 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543647
|
Invitrogen™
MA543647 |
20 μL |
Each for $213.50
|
|
|||||
Description
Neurogranin Monoclonal Antibody for Western Blot, IHC (P)
Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.Specifications
Neurogranin | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q92686 | |
4900 | |
Recombinant protein Neurogranin. The antigen corresponds to amino acid range 1-67 of the target protein. | |
Antibody |
C2 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
NRGN | |
0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; calmodulin-binding protein; hng; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); ng; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3; similar to neurogranin | |
NRGN | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title