Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurokinin B Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB30201A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30201A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30201A594 Supplier Novus Biologicals Supplier No. NB300201AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Neurokinin B Polyclonal specifically detects Neurokinin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Neurokinin B
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias gamma tachykinin 3, neurokinin beta, neurokinin B-like, neuromedin K, preprotachykinin B, PRO1155, tachykinin 3, tachykinin-3, ZNEUROK1NKNBNKB
Gene Symbols TAC3
Host Species Rabbit
Immunogen A synthetic peptide made to an internal sequence of mouse proNeurokinin B (∼93% homology to the rat sequence). [UniProtNo. P55099]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6866
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.