Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin 1/NLGN1 Antibody (S97A-31), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB242192488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB242192488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB242192488 Supplier Novus Biologicals Supplier No. NBP242192AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuroligin 1/NLGN1 Monoclonal specifically detects Neuroligin 1/NLGN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Neuroligin 1/NLGN1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S97A-31
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1
Gene Symbols NLGN1
Host Species Mouse
Immunogen Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin-2 and -3.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cognition and Behavior
Primary or Secondary Primary
Gene ID (Entrez) 22871
Test Specificity Greater then 40% identity with Neuroligin-2 and -3.Detects approx 120kDa. Does not cross-react with other Neuroligins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.