Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin 3/NLGN3 Antibody (242), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB068012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068012 Supplier Novus Biologicals Supplier No. NBP289824AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Neuroligin 3/NLGN3 Monoclonal specifically detects Neuroligin 3/NLGN3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Neuroligin 3/NLGN3
Applications Flow Cytometry
Classification Monoclonal
Clone 242
Conjugate Alexa Fluor 750
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Neuroligin 3/NLGN3 (Q9NZ94-2; Met1-Ser689).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54413
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.