Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuroligin 3/NLGN3 Antibody (S110-29), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB242200647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB242200647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB242200647 Supplier Novus Biologicals Supplier No. NBP242200AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuroligin 3/NLGN3 Monoclonal specifically detects Neuroligin 3/NLGN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Neuroligin 3/NLGN3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S110-29
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3
Gene Symbols NLGN3
Host Species Mouse
Immunogen Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3. Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ∼60% identity with Neuroligin-1. ∼40% identity with Neuroligin-2.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54413
Test Specificity Approx 60% identity with Neuroligin-1. approx 40% identity with Neuroligin-2.Detects approx 110kDa. Does not cross-react with Neuroligin-1, -2, -4 or -4.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.