Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuromedin BR/NMBR Antibody (466505), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB303441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303441 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303441 Supplier Novus Biologicals Supplier No. FAB4728N

Mouse Monoclonal Antibody

Neuromedin BR/NMBR Monoclonal antibody specifically detects Neuromedin BR/NMBR in Human samples. It is validated for Flow Cytometry, Flow Cytometry, CyTOF

Specifications

Antigen Neuromedin BR/NMBR
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 466505
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias neuromedin B receptor, neuromedin-B receptor, Neuromedin-B-preferring bombesin receptor, NMB-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NMBR, Met1-Met390, Accession # AAB27330
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4829
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.