Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin 1 Antibody (OTI5G9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP272965B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272965B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272965B Supplier Novus Biologicals Supplier No. NBP272965B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuronal Pentraxin 1 Monoclonal specifically detects Neuronal Pentraxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Neuronal Pentraxin 1
Applications Western Blot
Classification Monoclonal
Clone OTI5G9
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp686J2446, MGC105123, neuronal pentraxin INP1NP-I, neuronal pentraxin-1
Gene Symbols NPTX1
Host Species Mouse
Immunogen Full length recombinant protein of human Neuronal Pentraxin 1 (NP_002513) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4884.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.