Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Neuropathiazol
Click to view available options
Quantity:
10 mg
50 mg
Description
Neuropathiazol is a selective inducer of neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs). Selectively suppresses astrocyte differentiation when induced by LIF, BMP2 and FBS compared to retinoic acid. Enhances maturation of NPC derived neurons.
Specifications
Specifications
| Purity | >98% |
| Content And Storage | -20°C |
| Quantity | 10 mg |
| Product Type | Neuropathiazol |
| Molecular Weight (g/mol) | 338.42 |
| Inhibitors | Neuropathiazol |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction